2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 2-Hydroxy-4-diethylamino-2'-hydroxycarbonylbenzophenone, 2-(4-(Diethylamino)-2-hydroxybenzoyl)benzoic acid

D2140-25G D2140-100G
TCD2140-025GEA 51 USD
TCD2140-025G TCD2140-100G
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid ≥98.0% (by GC, titration analysis)
2-(4-Diethylamino-2-hydroxybenzoyl)benzoic acid

CAS Number: 5809-23-4
MDL Number: MFCD00134669
Molecular Formula: C18H19NO4
Molecular Weight: 313.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 180


Other Product Names / Synonyms: 2-Hydroxy-4-diethylamino-2'-hydroxycarbonylbenzophenone

Formula: C₁₈H₁₉NO₄
MW: 313.35 g/mol
Melting Pt: 180 °C
MDL Number: MFCD00134669
CAS Number: 5809-23-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=31ddaa649d1d6659f004228b2d2892d13201190e&vid=31ddaa649d1d6659f004228b2d2892d13201190e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR