(1S,2R)-2-(Dibutylamino)-1-phenyl-1-propanol ≥98.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
(-)-N,N-Dibutylnorephedrin, (-)-alpha-[1-(Dibutylamino)ethyl]benzyl Alcohol
CAS Number: 114389-70-7
MDL Number: MFCD00142695
Molecular Formula: C17H29NO
Molecular Weight: 263.43
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 170
Flash Point (°C): 110
Specific Gravity (20/20): 0.94
Specific rotation [a]20/D: -25 deg (C=2, Hexane)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks
Other Product Names / Synonyms: (-)-alpha-[1-(Dibutylamino)ethyl]benzyl Alcohol, (-)-N,N-Dibutylnorephedrin
Formula:
C₁₇H₂₉NO MW: 263.43 g/mol Boiling Pt: 170 °C Density: 0.94 (20 °C) Flash Pt: 110 °C |
MDL Number:
MFCD00142695 CAS Number: 114389-70-7 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...