1,2-Diamino-2-methylpropane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
2-Methyl-1,2-propanediamine, 2-Methylpropane-1,2-diamine
D2102-25ML
D2102-5ML
TCD2102-025MLEA
494
USD
TCD2102-025ML
TCD2102-5ML
1,2-Diamino-2-methylpropane ≥97.0% (by GC, titration analysis)
1,2-Diamino-2-methylpropane
CAS Number: 811-93-8
MDL Number: MFCD00008054
Molecular Formula: C4H12N2
Molecular Weight: 88.15
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 52
Flash Point (°C): 23
Specific Gravity (20/20): 0.85
Product Categories: Materials Chemistry, Monomers
Other Product Names / Synonyms: 2-Methyl-1,2-propanediamine
Formula:
C₄H₁₂N₂ MW: 88.15 g/mol Boiling Pt: 52 °C Density: 0.85 (20 °C) Flash Pt: 23 °C |
MDL Number:
MFCD00008054 CAS Number: 811-93-8 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...