Acrylamide ≥98.0% (by titrimetric analysis)

Supplier: TCI America
Danger

Synonyms: Prop-2-enamide

A0139-25G A0139-500G
TCA0139-025GEA 18.27 USD
TCA0139-025G TCA0139-500G
Acrylamide ≥98.0% (by titrimetric analysis)
Acrylamide (monomer)

CAS Number: 79-06-1
MDL Number: MFCD00008032
Molecular Formula: C3H5NO
Molecular Weight: 71.08
Purity/Analysis Method: 98.0% (T)
Form: Crystal
Boiling point (°C): 125
Melting point (°C): 85
Flash Point (°C): 138

Formula: CH₂=CHCONH₂
MW: 71.08 g/mol
Boiling Pt: 125 °C
Melting Pt: 85 °C
Density: 1.122 g/cm³ (20 °C)
Flash Pt: 138 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00008032
CAS Number: 79-06-1
EINECS: 201-173-7
UN: 2074
ADR: 6.1,III
https://pr.vwr.com/stibo/web/std.lang.all/75/84/28927584.jpg

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR