α,2,3-Trichlorotoluene ≥98.0%
Supplier: TCI America
Synonyms:
1,2-Dichloro-3-(chloromethyl)benzene, alpha,2,3-Trichlorotoluene, 1-(Chloromethyl)-2,3-dichlorobenzene, 2,3-Dichlorobenzyl chloride
D2060-5G
D2060-25G
TCD2060-005GEA
30
USD
TCD2060-005G
TCD2060-25G
α,2,3-Trichlorotoluene ≥98.0%
α,2,3-Trichlorotoluene
CAS Number: 3290-01-5
MDL Number: MFCD00035817
Molecular Formula: C7H5Cl3
Molecular Weight: 195.47
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 255
Specific Gravity (20/20): 1.43
Other Product Names / Synonyms: alpha,2,3-Trichlorotoluene
Formula:
C₇H₅Cl₃ MW: 195.47 g/mol Boiling Pt: 255 °C Density: 1.43 (20 °C) |
MDL Number:
MFCD00035817 CAS Number: 3290-01-5 EINECS: 221-947-8 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...