1,1-Diethylguanidine sulfate ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 1,1-Diethylguanidine hemisulphate

D2035-25G D2035-5G
TCD2035-025GEA 443 USD
TCD2035-025G TCD2035-5G
1,1-Diethylguanidine sulfate ≥98.0% (by titrimetric analysis)
1,1-Diethylguanidine sulfate

MDL Number: MFCD00035597
Molecular Formula: C5H13N3
Molecular Weight: 328.43
Purity/Analysis Method: >98.0% (T)
Form: Crystal

Formula: C₅H₁₃N₃
MW: 328.43 g/mol
MDL Number: MFCD00035597
CAS Number: 77297-00-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=116a0fb6ed1b832d231072409e020e64b378466d&vid=116a0fb6ed1b832d231072409e020e64b378466d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR