Diallyldimethylammonium chloride 60% in water
Supplier: TCI America
D2003-25ML
D2003-500ML
TCD2003-025MLEA
26.52
USD
TCD2003-025ML
TCD2003-500ML
Diallyldimethylammonium chloride 60% in water
Diallyldimethylammonium chloride
CAS Number: 7398-69-8
MDL Number: MFCD00043200
Molecular Formula: C8H16ClN
Molecular Weight: 161.67
Form: Clear Liquid
Color: Colorless
Formula:
C₈H₁₆ClN MW: 161.67 g/mol |
MDL Number:
MFCD00043200 CAS Number: 7398-69-8 |
|
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...