N-(3-Dimethylaminopropyl)methacrylamide ≥98.0% stabilised

Supplier: TCI America

Synonyms: N-(3-(Dimethylamino)propyl)methacrylamide

D1919-500G D1919-25G D1919-100G
TCD1919-500GEA 98.91 USD
TCD1919-500G TCD1919-025G TCD1919-100G
N-(3-Dimethylaminopropyl)methacrylamide ≥98.0% stabilised
N-(3-Dimethylaminopropyl)methacrylamide

(stabilized with MEHQ) CAS Number: 5205-93-6 MDL Number: MFCD00038359 Molecular Formula: C9H18N2O Molecular Weight: 170.26 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 130 Melting point (°C): -60 Flash Point (°C): 140 Specific Gravity (20/20): 0.95

Formula: C₉H₁₈N₂O
Boiling Pt: 130 °C
Melting Pt: –60 °C
Density: 0.95 (20 °C)
Flash Pt: 140 °C
MDL Number: MFCD00038359
CAS Number: 5205-93-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=05fece070f7cfb2517b73a3588c1a517e0dcec4e&vid=05fece070f7cfb2517b73a3588c1a517e0dcec4e&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR