D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
(1R,2R)-2-Amino-1-(4-nitrophenyl)propane-1,3-diol, (1R,2R)-(-)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
A1017-25G
A1017-500G
TCA1017-025GEA
80
USD
TCA1017-025G
TCA1017-500G
D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol ≥98.0% (by titrimetric analysis)
D-(-)-threo-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
CAS Number: 716-61-0
MDL Number: MFCD00078126
Molecular Formula: C9H12N2O4
Molecular Weight: 212.21
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 163
Specific rotation [a]20/D: -30.5 deg (C=1, 6mol/L HCl)
Product Categories: Synthetic Organic Chemistry, Optical Resolving Agents, Chiral Building Blocks
Other Product Names / Synonyms: (1R,2R)-(-)-2-Amino-1-(4-nitrophenyl)-1,3-propanediol
Formula:
C₉H₁₂N₂O₄ MW: 212.21 g/mol Melting Pt: 163 °C |
MDL Number:
MFCD00078126 CAS Number: 716-61-0 |
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...