3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate ≥98.0%

Supplier: TCI America

Synonyms: α-Methyl-L-β-3,4-Dihydroxyphenylalanine, alpha-Methyl-L-DOPA, Methyldopa

D1817-5G D1817-25G
TCD1817-5GEA 64.09 USD
TCD1817-5G TCD1817-025G
3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate ≥98.0%
3-(3,4-Dihydroxyphenyl)-2-methyl-L-alanine sesquihydrate

CAS Number: 555-30-6
MDL Number: MFCD00004186
Molecular Formula: C10H13NO4
Molecular Weight: 211.22
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Specific rotation [a]20/D: -14 deg (C=1, H2O)


Product Categories: Bioscience, Inhibitors (for Pharmacological Research), Neurotransmitter Agent Ingredients, Antihypertensive Ingredients, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: alpha-Methyl-L-DOPA, Methyldopa

Formula: C₁₀H₁₃NO₄
MW: 211.22 g/mol
MDL Number: MFCD00004186
CAS Number: 555-30-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c8eb17201860ea32a0e0b363911aa93dbd012e4c&vid=c8eb17201860ea32a0e0b363911aa93dbd012e4c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR