Diethylene glycol bis(2-chloroethyl)ether ≥97.0%

Supplier: TCI America

Synonyms: Diethylene Glycol Bis(2-chloroethyl) Ether, Bis[2-(2-chloroethoxy)ethyl] ether

D1784-25G D1784-500G
TCD1784-025GEA 101.72 USD
TCD1784-025G TCD1784-500G
Diethylene glycol bis(2-chloroethyl)ether ≥97.0%
Diethylene glycol bis(2-chloroethyl)ether

CAS Number: 638-56-2
MDL Number: MFCD00054473
Molecular Formula: C8H16Cl2O3
Molecular Weight: 231.11
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Flash Point (°C): 121
Specific Gravity (20/20): 1.18

Formula: C₈H₁₆Cl₂O₃
MW: 231.11 g/mol
Density: 1.18 (20 °C)
Flash Pt: 121 °C
MDL Number: MFCD00054473
CAS Number: 638-56-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a977931402d712844449f62c7c6866c566b9c103&vid=a977931402d712844449f62c7c6866c566b9c103&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR