1,3-Divinyltetramethyldisiloxane ≥98.0%

Supplier: TCI America

D1780-25ML D1780-100ML
TCD1780-025MLEA 23 USD
TCD1780-025ML TCD1780-100ML
1,3-Divinyltetramethyldisiloxane ≥98.0%
1,3-Divinyltetramethyldisiloxane

CAS Number: 2627-95-4
MDL Number: MFCD00014933
Molecular Formula: C8H18OSi2
Molecular Weight: 186.40
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 139
Flash Point (°C): 24
Specific Gravity (20/20): 0.81


Product Categories: Synthetic Organic Chemistry, Vinylation, Materials Chemistry, Monomers
Other Product Names / Synonyms: Tetramethyl-1,3-divinyldisiloxane

Formula: C₈H₁₈OSi₂
Boiling Pt: 139 °C
Density: 0.81 (20 °C)
Flash Pt: 24 °C
MDL Number: MFCD00014933
CAS Number: 2627-95-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=58acc4ed1d45921cefa5de664ae242009dcba835&vid=58acc4ed1d45921cefa5de664ae242009dcba835&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR