2,3-Dicyanopyrazine ≥99.0%
Supplier: TCI America
Synonyms:
Pyrazine-2,3-dicarbonitrile
D1751-25G
D1751-5G
D1751-100G
TCD1751-025GEA
104
USD
TCD1751-025G
TCD1751-005G
76218-982
2,3-Dicyanopyrazine ≥99.0%
2,3-Dicyanopyrazine
CAS Number: 13481-25-9
MDL Number: MFCD00059771
Molecular Formula: C6H2N4
Molecular Weight: 130.11
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 133
Product Categories: Materials Chemistry, Building Blocks for Phthalocyanines
Other Product Names / Synonyms: Pyrazine-2,3-dicarbonitrile
Formula:
C₆H₂N₄ MW: 130.1069 g/mol Melting Pt: 133 °C |
MDL Number:
MFCD00059771 CAS Number: 13481-25-9 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...