3-Dimethylamino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Dimethylaminoneopentanol

D1727-25ML D1727-500ML
TCD1727-025MLEA 100 USD
TCD1727-025ML TCD1727-500ML
3-Dimethylamino-2,2-dimethyl-1-propanol ≥97.0% (by GC, titration analysis)
3-Dimethylamino-2,2-dimethyl-1-propanol

CAS Number: 19059-68-8
MDL Number: MFCD00059879
Molecular Formula: C7H17NO
Molecular Weight: 131.22
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 69
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: Dimethylaminoneopentanol

Formula: C₇H₁₇NO
MW: 131.22 g/mol
Boiling Pt: 69 °C
Density: 0.86 (20 °C)
MDL Number: MFCD00059879
CAS Number: 19059-68-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d2e8bc1b44a1a12b861869311fd979f9d93b5fa8&vid=d2e8bc1b44a1a12b861869311fd979f9d93b5fa8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR