N,N-Dimethyl-n-octadecylamine ≥90.0%

Supplier: TCI America

Synonyms: N,N-Dimethyloctadecan-1-amine, Dymanthine, N,N-Dimethylstearylamine

D1609-25G D1609-250G
TCD1609-025GEA 45.7 USD
TCD1609-025G TCD1609-250G
N,N-Dimethyl-n-octadecylamine ≥90.0%
N,N-Dimethyl-n-octadecylamine

CAS Number: 124-28-7
MDL Number: MFCD00048496
Molecular Formula: C20H43N
Molecular Weight: 297.57
Purity/Analysis Method: >90.0% (GC)
Form: Crystal
Flash Point (°C): 163
Freezing point (°C): 21


Other Product Names / Synonyms: N,N-Dimethylstearylamine

Formula: C₂₀H₄₃N
MW: 297.57 g/mol
Flash Pt: 163 °C
MDL Number: MFCD00048496
CAS Number: 124-28-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=770dcf36195d87fea7e17bbefb139e62a9d3a144&vid=770dcf36195d87fea7e17bbefb139e62a9d3a144&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >90.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR