(R)-2,3-Diaminopropanoic acid hydrochloride ≥98.0% (by total nitrogen basis)
Supplier: TCI America
CAS Number: 6018-56-0
MDL Number: MFCD00070606
Molecular Formula: C3H8N2O2
Molecular Weight: 140.57
Purity/Analysis Method: >98.0% (N)
Form: Crystal
Melting point (°C): 240
Specific rotation [a]20/D: -23 deg (C=2, 1mol/L HCl)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: 3-Amino-D-alanine Hydrochloride
Formula:
C3H9ClN2O2 Melting Pt: 240 °C |
MDL Number:
MFCD00070606 CAS Number: 6018-56-0 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (N) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...