1,4-Dichloro-2-butyne ≥97.0%

Supplier: TCI America
Warning

Synonyms: 1,4-Dichlorobut-2-yne

D1528-25G D1528-5G
TCD1528-025GEA 156 USD
TCD1528-025G TCD1528-005G
1,4-Dichloro-2-butyne ≥97.0%
1,4-Dichloro-2-butyne

CAS Number: 821-10-3
MDL Number: MFCD00000981
Molecular Formula: C4H4Cl2
Molecular Weight: 122.98
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 166
Flash Point (°C): 160
Specific Gravity (20/20): 1.27
Storage Temperature: 0-10°C

Formula: C₄H₄Cl₂
MW: 122.98 g/mol
Boiling Pt: 166 °C
Melting Pt: –34.5 °C
Density: 1.27 (20 °C)
Flash Pt: 160 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00000981
CAS Number: 821-10-3
EINECS: 212-474-8
UN: 3334
ADR: 9,
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6b8c273c7e2e3ae03b8a39be507d45efda4fd40a&vid=6b8c273c7e2e3ae03b8a39be507d45efda4fd40a&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR