N-Butyldimethylamine ≥98.0%

Supplier: TCI America

Synonyms: N,N-Dimethylbutylamine, (Dimethylamino)butane

D1506-25ML D1506-500ML
TCD1506-025MLEA 46.79 USD
TCD1506-025ML TCD1506-500ML
N-Butyldimethylamine ≥98.0%
N-Butyldimethylamine

CAS Number: 927-62-8
MDL Number: MFCD00043838
Molecular Formula: C6H15N
Molecular Weight: 101.19
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 93
Flash Point (°C): -5
Specific Gravity (20/20): 0.72


Other Product Names / Synonyms: N,N-Dimethylbutylamine, (Dimethylamino)butane

Formula: C₆H₁₅N
MW: 101.19 g/mol
Boiling Pt: 93 °C
Density: 0.72 (20 °C)
Flash Pt: -5 °C
MDL Number: MFCD00043838
CAS Number: 927-62-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3342d75223edc2d574bf852f5e0a4c5c23e49023&vid=3342d75223edc2d574bf852f5e0a4c5c23e49023&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR