Dansyl-L-phenylalanine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dns-Phe-OH, 5-Dimethylaminonaphthalene-1-sulphonyl-L-phenylalanine

D1500-5G D1500-1G D1500-100MG
TCD1500-5GEA 442 USD
TCD1500-5G TCD1500-001G TCD1500-100MG
Dansyl-L-phenylalanine ≥98.0% (by titrimetric analysis)
Dansyl-L-phenylalanine

CAS Number: 1104-36-5
MDL Number: MFCD00037730
Molecular Formula: C21H22N2O4S
Molecular Weight: 398.48
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 172


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-Dimethylaminonaphthalene-1-sulfonyl-L-phenylalanine, Dns-Phe-OH

Formula: C₂₁H₂₂N₂O₄S
MW: 398.48 g/mol
Melting Pt: 172 °C
MDL Number: MFCD00037730
CAS Number: 1104-36-5
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fbecc24e9df32c6cf9944649a616e80ea270dafd&vid=fbecc24e9df32c6cf9944649a616e80ea270dafd&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR