Dansyl-L-leucine ≥98.0%

Supplier: TCI America

Synonyms: 5-Dimethylaminonaphthalene-1-sulphonyl-L-leucine, Dns-Leu-OH

D1498-1G D1498-100MG
TCD1498-001GEA 172 USD
TCD1498-001G TCD1498-100MG
Dansyl-L-leucine ≥98.0%
Dansyl-L-leucine

CAS Number: 1100-22-7
MDL Number: MFCD00069532
Molecular Formula: C18H24N2O4S
Molecular Weight: 364.46
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 126
Specific rotation [a]20/D: -9.5 deg (C=1, EtOH)


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: 5-Dimethylaminonaphthalene-1-sulfonyl-L-leucine, Dns-Leu-OH

Formula: C₁₈H₂₄N₂O₄S
MW: 364.46 g/mol
Melting Pt: 126 °C
MDL Number: MFCD00069532
CAS Number: 1100-22-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=a8385f0d18f72c4f9a97fc0916f85f50a2a3543b&vid=a8385f0d18f72c4f9a97fc0916f85f50a2a3543b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR