3'-(Dimethylamino)acetophenone ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1446-25G D1446-5G
TCD1446-025GEA 604 USD
TCD1446-025G TCD1446-005G
3'-(Dimethylamino)acetophenone ≥98.0% (by GC, titration analysis)
3'-(Dimethylamino)acetophenone

CAS Number: 18992-80-8
MDL Number: MFCD00059343
Molecular Formula: C10H13NO
Molecular Weight: 163.22
Purity/Analysis Method: >98.0% (GC,T)
Form: Crystal
Melting point (°C): 37

Formula: C₁₀H₁₃NO
MW: 163.216 g/mol
Melting Pt: 37 °C
MDL Number: MFCD00059343
CAS Number: 18992-80-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=0e9a172c9c6600434095338b05b98f42250e548f&vid=0e9a172c9c6600434095338b05b98f42250e548f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR