N,N-Diethylglycine methyl ester ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D1443-500G D1443-25G
TCD1443-500GEA 692.6 USD
TCD1443-500G TCD1443-025G
N,N-Diethylglycine methyl ester ≥98.0% (by GC, titration analysis)
N,N-Diethylglycine methyl ester

CAS Number: 30280-35-4 MDL Number: MFCD00059368 Molecular Formula: C7H15NO2 Molecular Weight: 145.20 Purity/Analysis Method: 98.0% (GC,T) Form: Clear Liquid Boiling point (°C): 86 Specific Gravity (20/20): 0.93 Storage Temperature: 0-10°C

Formula: C₇H₁₅NO₂
Boiling Pt: 86 °C
Density: 0.93 (20 °C)
MDL Number: MFCD00059368
CAS Number: 30280-35-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=306bf01d3adee6577aff04054b98fd6b8137ec8b&vid=306bf01d3adee6577aff04054b98fd6b8137ec8b&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR