Methyl dihydrojasmonate (cis and trans mixture) ≥95.0%

Supplier: TCI America

D1431-25ML D1431-100ML
TCD1431-025MLEA 52 USD
TCD1431-025ML TCD1431-100ML
Methyl dihydrojasmonate (cis and trans mixture) ≥95.0%
Methyl dihydrojasmonate (cis and trans mixture)

CAS Number: 24851-98-7
MDL Number: MFCD00151188
Molecular Formula: C13H22O3
Molecular Weight: 226.32
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 110
Flash Point (°C): 156
Specific Gravity (20/20): 1.00


Product Categories: Bioscience, Jasmonates
Other Product Names / Synonyms: 2-Amylcyclopentan-1-one-3-acetic Acid Methyl Ester, Dihydrojasmonic Acid Methyl Ester, Methyl 3-Oxo-2-pentyl-1-cyclopentaneacetate

Formula: C₁₃H₂₂O₃
Boiling Pt: 110 °C
Density: 1.00 (20 °C)
Flash Pt: 156 °C
MDL Number: MFCD00151188
CAS Number: 24851-98-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=3a46518775f4e889b19668306c361211627f57cc&vid=3a46518775f4e889b19668306c361211627f57cc&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR