Dicyclohexylamine nitrite ≥97.0%
Supplier: TCI America
Synonyms:
N,N-Dicyclohexylammonium nitrite
D1392-500G
D1392-25G
TCD1392-500GEA
187
USD
TCD1392-500G
TCD1392-025G
Dicyclohexylamine nitrite ≥97.0%
Dicyclohexylamine nitrite
CAS Number: 3129-91-7
MDL Number: MFCD00013128
Molecular Formula: C12H23N
Molecular Weight: 228.34
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 183
Formula:
C₁₂H₂₃N MW: 228.34 g/mol Melting Pt: 183 °C |
MDL Number:
MFCD00013128 CAS Number: 3129-91-7 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...