5-Phenyl-o-anisidine ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: 2-Amino-4-phenylanisole, 2-Methoxy-5-phenylaniline, 5-Phenyl-o-anisidine, 3-Amino-4-methoxybiphenyl

A0958-5G A0958-25G
TCA0958-005GEA 114 USD
TCA0958-005G TCA0958-025G
5-Phenyl-o-anisidine ≥98.0% (by titrimetric analysis)
5-Phenyl-o-anisidine

CAS Number: 39811-17-1
MDL Number: MFCD00007790
Molecular Formula: C13H13NO
Molecular Weight: 199.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Melting point (°C): 84


Other Product Names / Synonyms: 2-Amino-4-phenylanisole, 5-Phenyl-o-anisidine

Formula: C₁₃H₁₃NO
MW: 199.25 g/mol
Melting Pt: 84 °C
MDL Number: MFCD00007790
CAS Number: 39811-17-1
EINECS: 254-639-7
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7720f1e9770633c6cc3e7c2d26965b01ea1ca015&vid=7720f1e9770633c6cc3e7c2d26965b01ea1ca015&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR