(S)-2,3-Diaminopropanoic acid hydrochloride ≥97.0% (by titrimetric analysis)
Supplier: TCI America
CAS Number: 1482-97-9
MDL Number: MFCD00065497
Molecular Formula: C3H8N2O2
Molecular Weight: 140.57
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 233
Specific rotation [a]20/D: 24 deg (C=2, 1mol/L HCl)
Product Categories: Synthetic Organic Chemistry, Chiral Building Blocks, Bioscience, non-Proteinogenic Amino Acids
Other Product Names / Synonyms: L-3-Aminoalanine Hydrochloride
Formula:
C₃H₈N₂O₂·HCl Melting Pt: 233 °C |
MDL Number:
MFCD00065497 CAS Number: 1482-97-9 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...