Clonidine hydrochloride ≥98.0%

Supplier: TCI America

Synonyms: 2-(2,6-Dichlorophenylamino)-2-imidazoline Hydrochloride, N-(2,6-Dichlorophenyl)-4,5-dihydro-1H-imidazol-2-amine hydrochloride, 2-(2,6-Dichloroanilino)-2-imidazoline Hydrochloride

D1353-1G D1353-5G
TCD1353-001GEA 74 USD
TCD1353-001G TCD1353-005G
Clonidine hydrochloride ≥98.0%
Clonidine hydrochloride

CAS Number: 4205-91-8
MDL Number: MFCD00036705
Molecular Formula: C9H9Cl2N3
Molecular Weight: 266.55
Purity/Analysis Method: >98.0% (HPLC,T)
Form: Crystal
Melting point (°C): 312


Product Categories: Bioscience, Antihypertensive Ingredients
Other Product Names / Synonyms: Clonidine Hydrochloride, 2-(2,6-Dichlorophenylamino)-2-imidazoline Hydrochloride

Formula: C₉H₉Cl₂N₃
MW: 266.55 g/mol
Melting Pt: 312 °C
MDL Number: MFCD00036705
CAS Number: 4205-91-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e4bf3b9cb962e7a5f6e94dd8e082ec18518c139d&vid=e4bf3b9cb962e7a5f6e94dd8e082ec18518c139d&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (HPLC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR