5,5-Diphenylhydantoin sodium salt ≥98.0% (by titrimetric analysis)
Supplier: TCI America
Synonyms:
Phenytoin sodium, Sodium 5,5-Diphenylhydantoin, Sodium 2,4-dioxo-5,5-diphenylimidazolidin-1-ide
D1331-25G
D1331-500G
TCD1331-025GEA
33
USD
TCD1331-025G
TCD1331-500G
5,5-Diphenylhydantoin sodium salt ≥98.0% (by titrimetric analysis)
5,5-Diphenylhydantoin sodium salt
CAS Number: 630-93-3
MDL Number: MFCD00069674
Molecular Formula: C15H12N2O2
Molecular Weight: 274.25
Purity/Analysis Method: >98.0% (T)
Form: Crystal
Other Product Names / Synonyms: Sodium 5,5-Diphenylhydantoin
Formula:
C₁₅H₁₂N₂O₂ MW: 274.25 g/mol |
MDL Number:
MFCD00069674 CAS Number: 630-93-3 |
|
Specification Test Results
Purity/Analysis Method | >98.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...