Decamethonium iodide
Supplier: TCI America
Synonyms:
Decane-1,10-bis(trimethylammonium Iodide), Decamethylenebis(trimethylammonium Iodide)
D1315-5G
D1315-25G
TCD1315-005GEA
77
USD
TCD1315-005G
TCD1315-025G
Decamethonium iodide
Decamethonium iodide
CAS Number: 1420-40-2
MDL Number: MFCD00031553
Molecular Formula: C16H38I2N2
Molecular Weight: 512.30
Form: Crystal
Color: White
Other Product Names / Synonyms: Decamethylenebis(trimethylammonium Iodide), Decane-1,10-bis(trimethylammonium Iodide)
Formula:
C₁₆H₃₈I₂N₂ MW: 512.30 g/mol |
MDL Number:
MFCD00031553 CAS Number: 1420-40-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...