Decamethonium bromide ≥98.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Decane-1,10-bis(trimethylammonium Bromide), Decamethonium Bromide, Decamethylenebis(trimethylammonium Bromide)

D1314-25G D1314-5G
TCD1314-025GEA 293 USD
TCD1314-025G TCD1314-005G
Decamethonium bromide ≥98.0% (by titrimetric analysis)
Decamethonium bromide

CAS Number: 541-22-0
MDL Number: MFCD00011779
Molecular Formula: C16H38Br2N2
Molecular Weight: 418.30
Purity/Analysis Method: >98.0% (T)
Form: Crystal


Other Product Names / Synonyms: Decamethylenebis(trimethylammonium Bromide), Decane-1,10-bis(trimethylammonium Bromide)

Formula: C₁₆H₃₈Br₂N₂
MW: 418.30 g/mol
MDL Number: MFCD00011779
CAS Number: 541-22-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=57868f9b896b2074994eccb1a51fa9bc2b10e3ea&vid=57868f9b896b2074994eccb1a51fa9bc2b10e3ea&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR