N,N'-Diethyl-1,6-diaminohexane ≥97.0% (by GC, titration analysis)
Supplier: TCI America
Synonyms:
N1,N6-Diethylhexane-1,6-diamine
D1305-25ML
D1305-5ML
TCD1305-025MLEA
416.4
USD
TCD1305-025ML
TCD1305-5ML
N,N'-Diethyl-1,6-diaminohexane ≥97.0% (by GC, titration analysis)
N,N'-Diethyl-1,6-diaminohexane
CAS Number: 13093-05-5
MDL Number: MFCD00026707
Molecular Formula: C10H24N2
Molecular Weight: 172.32
Purity/Analysis Method: >97.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 228
Specific Gravity (20/20): 0.83
Other Product Names / Synonyms: 1,6-Bis(ethylamino)hexane, N,N'-Diethylhexamethylenediamine, N,N'-Diethyl-1,6-hexanediamine
Formula:
C₁₀H₂₄N₂ MW: 172.3110 g/mol Boiling Pt: 228 °C Density: 0.83 (20 °C) |
MDL Number:
MFCD00026707 CAS Number: 13093-05-5 |
Specification Test Results
Purity/Analysis Method | >97.0% (GC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...