N,N-Dimethylformamide dibutyl acetal ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: 1,1-Dibutoxytrimethylamine

D1302-5ML D1302-25ML
TCD1302-005MLEA 47 USD
TCD1302-005ML TCD1302-025ML
N,N-Dimethylformamide dibutyl acetal ≥98.0% (by GC, titration analysis)
N,N-Dimethylformamide dibutyl acetal

[for Esterification]
CAS Number: 18503-90-7
MDL Number: MFCD00015249
Molecular Formula: C11H25NO2
Molecular Weight: 203.33
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 98
Flash Point (°C): 53
Specific Gravity (20/20): 0.86


Product Categories: Synthetic Organic Chemistry, Alkylation (Esterification), Analytical Science, Esterification & Alkylation (GC Derivatizing Reagents)
Other Product Names / Synonyms: 1,1-Dibutoxytrimethylamine

Formula: C₁₁H₂₅NO₂
MW: 203.32 g/mol
Boiling Pt: 98 °C
Density: 0.86 (20 °C)
Flash Pt: 53 °C
MDL Number: MFCD00015249
CAS Number: 18503-90-7
EINECS: 242-387-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78703f4e7e905133a15a4c9f1fcb78862eacfe58&vid=0dfc300e0f3da5d374e4f51e8738b061f6fa2e72&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR