N,N'-Dimethyl-1,6-diaminohexane ≥97.0%

Supplier: TCI America

Synonyms: N1,N6-Dimethylhexane-1,6-diamine

D1298-10ML D1298-25ML
TCD1298-010MLEA 169.48 USD
TCD1298-010ML TCD1298-25ML
N,N'-Dimethyl-1,6-diaminohexane ≥97.0%
N,N'-Dimethyl-1,6-diaminohexane

CAS Number: 13093-04-4
MDL Number: MFCD00008293
Molecular Formula: C8H20N2
Molecular Weight: 144.26
Purity/Analysis Method: >97.0% (GC)
Form: Clear Liquid
Boiling point (°C): 205
Flash Point (°C): 112
Freezing point (°C): 18
Specific Gravity (20/20): 0.83


Other Product Names / Synonyms: 1,6-Bis(methylamino)hexane, N,N'-Dimethyl-1,6-hexanediamine

Formula: C₈H₂₀N₂
MW: 144.2578 g/mol
Boiling Pt: 205 °C
Density: 0.83 (20 °C)
Flash Pt: 112 °C
MDL Number: MFCD00008293
CAS Number: 13093-04-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6a9882a01ca23fa133ab9831efafd144f48f23f8&vid=6a9882a01ca23fa133ab9831efafd144f48f23f8&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR