3,5-Dimethylcyclohexanone ≥98.0%
Supplier: TCI America
D1282-25ML
D1282-5ML
TCD1282-025MLEA
233.3
USD
TCD1282-025ML
TCD1282-005ML
3,5-Dimethylcyclohexanone ≥98.0%
3,5-Dimethylcyclohexanone
CAS Number: 2320-30-1
MDL Number: MFCD00019467
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >98.0% (GC)
Form: Clear Liquid
Boiling point (°C): 176
Specific Gravity (20/20): 0.89
Formula:
C₈H₁₄O MW: 126.1962 g/mol Boiling Pt: 176 °C Density: 0.89 (20 °C) |
MDL Number:
MFCD00019467 CAS Number: 2320-30-1 |
Specification Test Results
Purity/Analysis Method | >98.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...