2,5-Dimethylcyclohexanone ≥95.0%

Supplier: TCI America

D1279-25ML D1279-5ML
TCD1279-025MLEA 734 USD
TCD1279-025ML TCD1279-5ML
2,5-Dimethylcyclohexanone ≥95.0%
2,5-Dimethylcyclohexanone

CAS Number: 932-51-4
MDL Number: MFCD00045551
Molecular Formula: C8H14O
Molecular Weight: 126.20
Purity/Analysis Method: >95.0% (GC)
Form: Clear Liquid
Boiling point (°C): 80
Flash Point (°C): 51
Specific Gravity (20/20): 0.91
Storage Temperature: 0-10°C

Formula: C₈H₁₄O
MW: 126.2 g/mol
Boiling Pt: 80 °C
Density: 0.91 (20 °C)
Flash Pt: 51 °C
MDL Number: MFCD00045551
CAS Number: 932-51-4
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=663b11fc6c7d710e10df340d68b7f9aea571c47b&vid=120d837dc75867773b7a75dbec86ecc0a35c346f&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >95.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR