Ferbam (irontris(dimethyldithiocarbamate)) ≥97.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Ferric dimethyldithiocarbamate, Ferbam, Iron(III) dimethyldithiocarbamate, Dimethyldithiocarbamic acid Ferric salt

D1267-500G D1267-25G
TCD1267-500GEA 201 USD
TCD1267-500G TCD1267-025G
Ferbam (irontris(dimethyldithiocarbamate)) ≥97.0% (by titrimetric analysis)
Ferbam (irontris(dimethyldithiocarbamate))

CAS Number: 14484-64-1
MDL Number: MFCD00067269
Molecular Formula: C3H7NS2
Molecular Weight: 416.47
Purity/Analysis Method: >97.0% (T)
Form: Crystal


Product Categories: Materials Chemistry, Magnetic Metal Complexes (Magnetic Materials)
Other Product Names / Synonyms: Dimethyldithiocarbamic Acid Ferric Salt, Ferbam, Ferric Dimethyldithiocarbamate

Formula: C₉H₁₈FeN₃S₆
MDL Number: MFCD00067269
CAS Number: 14484-64-1
EINECS: 238-484-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=4665205e495d535816276c2f0848c899abcdf7c7&vid=4665205e495d535816276c2f0848c899abcdf7c7&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >97.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR