2,4-Dimethyl-1-heptene ≥98.0%

Supplier: TCI America

D1258-5ML D1258-1ML
TCD1258-005MLEA 163 USD
TCD1258-005ML TCD1258-001ML
2,4-Dimethyl-1-heptene ≥98.0%
2,4-Dimethyl-1-heptene

CAS Number: 19549-87-2 MDL Number: MFCD00059250 Molecular Formula: C9H18 Molecular Weight: 126.24 Purity/Analysis Method: 98.0% (GC) Form: Clear Liquid Boiling point (°C): 134 Specific Gravity (20/20): 0.73

Formula: C₉H₁₈
MW: 126.24 g/mol
Boiling Pt: 134 °C
Density: 0.73 (20 °C)
MDL Number: MFCD00059250
CAS Number: 19549-87-2

Order Now

Specification Test Results

Purity/Analysis Method 98.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR