Sodium dodecylbenzenesulfonate (mixture) 62,0% wetted with water, soft type
Supplier: TCI America
Synonyms:
Dodecylbenzenesulphonic acid Sodium Salt, Laurylbenzenesulphonic acid Sodium Salt, Alkylbenzenesulphonic acid Sodium Salt, Sodium Alkylbenzenesulphonate, Sodium Laurylbenzenesulphonate
CAS Number: 69669-44-9
MDL Number: MFCD00011508
Molecular Formula: C18H30O3S
Molecular Weight: 348.48
Form: Paste
Color: Very Pale Yellow
Flash Point (°C): 110
Other Product Names / Synonyms: Dodecylbenzenesulfonic Acid Sodium Salt, Laurylbenzenesulfonic Acid Sodium Salt, Sodium Laurylbenzenesulfonate, Alkylbenzenesulfonic Acid Sodium Salt, Sodium Alkylbenzenesulfonate
Formula:
C₁₈H₃₀O₃S MW: 348.48 g/mol Flash Pt: 110 °C |
MDL Number:
MFCD00011508 CAS Number: 69669-44-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...