cis-2,6-Dimethylpiperidine 99,0

Supplier: TCI America

Synonyms: cis-2,6-Lupetidine

D1198-25ML D1198-500ML
TCD1198-025MLEA 38.89 USD
TCD1198-025ML TCD1198-500ML
cis-2,6-Dimethylpiperidine 99,0
cis-2,6-Dimethylpiperidine

CAS Number: 766-17-6
MDL Number: MFCD00066304
Molecular Formula: C7H15N
Molecular Weight: 113.20
Purity/Analysis Method: >99.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 127
Flash Point (°C): 12
Specific Gravity (20/20): 0.82


Other Product Names / Synonyms: cis-Hexahydro-2,6-lutidine, cis-Lupetidine

Formula: C₇H₁₅N
MW: 113.20 g/mol
Boiling Pt: 127 °C
Density: 0.82 (20 °C)
Flash Pt: 12 °C
MDL Number: MFCD00066304
CAS Number: 766-17-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f46df621dbd8b54f2a4de2afe0b4559157712a31&vid=f46df621dbd8b54f2a4de2afe0b4559157712a31&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR