2,4-Dihydroxy-5,6-dimethylpyrimidine ≥99.0% (by HPLC, titration analysis)
Supplier: TCI America
Synonyms:
5,6-Dimethyl-2,4-pyrimidinediol, 5,6-Dimethyluracil, 2,4-Dihydroxy-5,6-dimethylpyrimidine
D1136-1G
D1136-5G
TCD1136-001GEA
69
USD
TCD1136-001G
TCD1136-005G
2,4-Dihydroxy-5,6-dimethylpyrimidine ≥99.0% (by HPLC, titration analysis)
2,4-Dihydroxy-5,6-dimethylpyrimidine
CAS Number: 26305-13-5
MDL Number: MFCD00010708
Molecular Formula: C6H8N2O2
Molecular Weight: 140.14
Purity/Analysis Method: >99.0% (HPLC,T)
Form: Crystal
Other Product Names / Synonyms: 2,4-Dihydroxy-5,6-dimethylpyrimidine, 5,6-Dimethyl-2,4-pyrimidinediol
Formula:
C₆H₈N₂O₂ MW: 140.1400 g/mol |
MDL Number:
MFCD00010708 CAS Number: 26305-13-5 |
Specification Test Results
Purity/Analysis Method | >99.0% (HPLC,T) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...