3,5-Diiodo-L-thyronine ≥96.0%
Supplier: TCI America
Synonyms:
3,5-Diiodo-L-thyronine, (S)-2-Amino-3-(4-(4-hydroxyphenoxy)-3,5-diiodophenyl)propanoic acid
D1119-1G
D1119-100MG
TCD1119-1GEA
236.31
USD
TCD1119-1G
TCD1119-100MG
3,5-Diiodo-L-thyronine ≥96.0%
3,5-Diiodo-L-thyronine
CAS Number: 1041-01-6
MDL Number: MFCD00064987
Molecular Formula: C15H13I2NO4
Molecular Weight: 525.08
Purity/Analysis Method: >96.0% (T)
Form: Crystal
Melting point (°C): 256
Specific rotation [a]20/D: 29 deg (C=1, 1mol/L HCl:EtOH=1:2)
Formula:
C₁₅H₁₃I₂NO₄ MW: 525.08 g/mol Melting Pt: 256 °C |
MDL Number:
MFCD00064987 CAS Number: 1041-01-6 |
|
Specification Test Results
Purity/Analysis Method | >96.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...