2-Nitro-p-xylene ≥99.0%
Supplier: TCI America
Synonyms:
2,5-Dimethylnitrobenzene, Nitro-p-xylene, 1,4-Dimethyl-2-nitrobenzene
D1105-25G
D1105-500G
TCD1105-025GEA
31
USD
TCD1105-025G
TCD1105-500G
2-Nitro-p-xylene ≥99.0%
2-Nitro-p-xylene
CAS Number: 89-58-7
MDL Number: MFCD00024284
Molecular Formula: C8H9NO2
Molecular Weight: 151.17
Purity/Analysis Method: >99.0% (GC)
Form: Clear Liquid
Boiling point (°C): 104
Flash Point (°C): 104
Specific Gravity (20/20): 1.13
Other Product Names / Synonyms: Nitro-p-xylene
Formula:
C₈H₉NO₂ MW: 151.16 g/mol Boiling Pt: 104 °C Melting Pt: –25 °C Density: 1.13 (20 °C) Flash Pt: 104 °C |
MDL Number:
MFCD00024284 CAS Number: 89-58-7 EINECS: 201-920-7 |
Specification Test Results
Purity/Analysis Method | >99.0% (GC) |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...