N,N-Dimethylacrylamide ≥99.0% stabilized
Supplier: TCI America
(stabilized with MEHQ)
CAS Number: 2680-03-7
MDL Number: MFCD00008626
Molecular Formula: C5H9NO
Molecular Weight: 99.13
Purity/Analysis Method:
Formula:
C₅H₉NO Boiling Pt: 80 °C Density: 0.97 (20 °C) Flash Pt: 77 °C |
MDL Number:
MFCD00008626 CAS Number: 2680-03-7 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...