N-(2,4-Dinitrophenyl)-L-leucine ≥99.0% (by titrimetric analysis)

Supplier: TCI America

Synonyms: Dnp-L-Leucine, Dnp-Leu-OH, N-Dnp-L-leucine

D1033-100MG D1033-1G
TCD1033-100MGEA 39 USD
TCD1033-100MG TCD1033-001G
N-(2,4-Dinitrophenyl)-L-leucine ≥99.0% (by titrimetric analysis)
N-(2,4-Dinitrophenyl)-L-leucine

CAS Number: 1655-57-8
MDL Number: MFCD00038102
Molecular Formula: C12H15N3O6
Molecular Weight: 297.27
Purity/Analysis Method: >99.0% (T)
Form: Crystal
Melting point (°C): 98
Specific rotation [a]20/D: 56 deg (C=0.5, NaHCO3 aq.(4%))


Product Categories: Bioscience, Amino Acids (N-Protected)
Other Product Names / Synonyms: N-Dnp-L-leucine, Dnp-Leu-OH

Formula: C₁₂H₁₅N₃O₆
MW: 297.27 g/mol
Melting Pt: 98 °C
MDL Number: MFCD00038102
CAS Number: 1655-57-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=c5911b2f6197ce87b9662017f0b24bcd3cbf8fa9&vid=c5911b2f6197ce87b9662017f0b24bcd3cbf8fa9&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR