2-(Dimethylamino)ethyl acetate ≥98.0% (by GC, titration analysis)

Supplier: TCI America

Synonyms: Acetic Acid 2-(Dimethylamino)ethyl Ester

A0929-25ML A0929-500ML
TCA0929-025MLEA 81 USD
TCA0929-025ML TCA0929-500ML
2-(Dimethylamino)ethyl acetate ≥98.0% (by GC, titration analysis)
2-(Dimethylamino)ethyl acetate

CAS Number: 1421-89-2
MDL Number: MFCD00041912
Molecular Formula: C6H13NO2
Molecular Weight: 131.18
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 152
Specific Gravity (20/20): 0.93


Other Product Names / Synonyms: Acetic Acid 2-(Dimethylamino)ethyl Ester

Formula: C₆H₁₃NO₂
MW: 131.18 g/mol
Boiling Pt: 152 °C
Density: 0.93 (20 °C)
MDL Number: MFCD00041912
CAS Number: 1421-89-2
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=78de56530f5617ce0e6a5b5aae48086faed6ccc6&vid=78de56530f5617ce0e6a5b5aae48086faed6ccc6&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR