tert-Dodecyl mercaptan (mixture of isomers) ≥98.0%

Supplier: TCI America

Synonyms: tert-Lauryl Mercaptan, tert-Dodecanethiol, Tert-dodecylmercaptan

D0971-25ML D0971-500ML
TCD0971-025MLEA 40.38 USD
TCD0971-025ML TCD0971-500ML
tert-Dodecyl mercaptan (mixture of isomers) ≥98.0%
tert-Dodecyl mercaptan (mixture of isomers)

CAS Number: 25103-58-6
MDL Number: MFCD00043233
Molecular Formula: C12H26S
Molecular Weight: 202.40
Purity/Analysis Method: >98.0% (T)
Form: Clear Liquid
Flash Point (°C): 110
Specific Gravity (20/20): 0.86


Other Product Names / Synonyms: tert-Dodecanethiol, tert-Lauryl Mercaptan

Formula: C₁₂H₂₆S
MW: 202.40 g/mol
Density: 0.86 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00043233
CAS Number: 25103-58-6

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR