2,2'-Dithiodipropionic acid ≥98.0%
Supplier: TCI America
D0946-25G
D0946-5G
TCD0946-025GEA
469
USD
TCD0946-025G
TCD0946-5G
2,2'-Dithiodipropionic acid ≥98.0%
2,2'-Dithiodipropionic acid
CAS Number: 4775-93-3
MDL Number: MFCD00059649
Molecular Formula: C6H10O4S2
Molecular Weight: 210.26
Purity/Analysis Method:
Formula:
C₆H₁₀O₄S₂ |
MDL Number:
MFCD00059649 CAS Number: 4775-93-3 |
|
Specification Test Results
Purity/Analysis Method |
|
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...