Bis(4-nitrophenyl)sulfide ≥99.0%

Supplier: TCI America

Synonyms: 4,4'-Dinitrodiphenyl Sulphide, Di(4-nitrophenyl) sulphide

D0847-5G D0847-25G
TCD0847-005GEA 63.8 USD
TCD0847-005G TCD0847-025G
Bis(4-nitrophenyl)sulfide ≥99.0%
Bis(4-nitrophenyl)sulfide

CAS Number: 1223-31-0
MDL Number: MFCD00039745
Molecular Formula: C12H8N2O4S
Molecular Weight: 276.27
Purity/Analysis Method: >99.0% (GC)
Form: Crystal
Melting point (°C): 160


Product Categories: Materials Chemistry, Diphenyl Sulfides (for High-Performance Polymer Research)
Other Product Names / Synonyms: 4,4'-Dinitrodiphenyl Sulfide, Di(4-nitrophenyl) Sulfide

Formula: C₁₂H₈N₂O₄S
MW: 276.27 g/mol
Melting Pt: 160 °C
MDL Number: MFCD00039745
CAS Number: 1223-31-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=f60a685cc4c195110fe5bb28728bf2e4bb169f6c&vid=f60a685cc4c195110fe5bb28728bf2e4bb169f6c&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >99.0% (GC)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR