N-Methyldicyclohexylamine ≥98.0% (by GC, titration analysis)

Supplier: TCI America

D0820-25ML D0820-500ML
TCD0820-025MLEA 23.4 USD
TCD0820-025ML TCD0820-500ML
N-Methyldicyclohexylamine ≥98.0% (by GC, titration analysis)
N-Methyldicyclohexylamine

CAS Number: 7560-83-0
MDL Number: MFCD00014289
Molecular Formula: C13H25N
Molecular Weight: 195.35
Purity/Analysis Method: >98.0% (GC,T)
Form: Clear Liquid
Boiling point (°C): 150
Flash Point (°C): 110
Specific Gravity (20/20): 0.92


Other Product Names / Synonyms: N-Methyldicyclohexylamine

Formula: C₁₃H₂₅N
Boiling Pt: 150 °C
Density: 0.92 (20 °C)
Flash Pt: 110 °C
MDL Number: MFCD00014289
CAS Number: 7560-83-0
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=ab4d61d6cfd11aa856c34a170dc71d4917cfe776&vid=ab4d61d6cfd11aa856c34a170dc71d4917cfe776&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Purity/Analysis Method >98.0% (GC,T)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR