1,3-Dimethyl-2-thiourea ≥97.0% for synthesis
Supplier: TCI America
Synonyms:
1,3-Dimethylthiourea, N,N'-Dimethylthiourea
D0804-25G
D0804-500G
D0804-100G
TCD0804-025GEA
92
USD
TCD0804-025G
TCD0804-500G
76218-962
1,3-Dimethyl-2-thiourea ≥97.0% for synthesis
1,3-Dimethyl-2-thiourea
CAS Number: 534-13-4
MDL Number: MFCD00004923
Molecular Formula: C3H8N2S
Molecular Weight: 104.17
Purity/Analysis Method: >97.0% (T)
Form: Crystal
Melting point (°C): 65
Formula:
C₃H₈N₂S MW: 104.18 g/mol Boiling Pt: 133 Melting Pt: 65 °C |
MDL Number:
MFCD00004923 CAS Number: 534-13-4 EINECS: 208-588-2 Merck Index: 13,03287 |
|
Specification Test Results
Purity/Analysis Method | >97.0% (T) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...